Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric acid, Puriss. p.a., crystallized, ≥99.0% (T), Solstice

Catalog No. 60047635 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-635 500 g Glass Bottle
60-011-38 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-635 Supplier Solstice Supplier No. 79622500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., crystallized,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Melting Point ∼40°C (literature)
Boiling Point 158°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Density 1.685 g/mL at 25 °C(lit.)
Packaging Glass Bottle
Vapor Pressure 2.2 mmHg (20°C), 5 mmHg (25°C)
Quantity 500 g
Linear Formula H3PO4
UN Number UN3453
Beilstein 1921286
Formula Weight 98
Percent Purity ≥99%
Grade puriss. p.a.
Chemical Name or Material Phosphoric acid, Puriss. p.a., crystallized, ≥99.0% (T)
Show More Show Less
Hazard Statement P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
ShelfLife 1620 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Phosphoric acid, solid
EINECSNumber 231-633-2
RTECSNumber 231-633-2
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.