Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorous-32, MP Biomedicals™

Catalog No. MP164014L02 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
2mCi; 74MBq
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP164014L02 2mCi; 74MBq
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP164014L02 Supplier MP Biomedicals, Inc Supplier No. 0164014L02

Chemical Identifiers

Molecular Formula H3PO4 in H2O, HCl free

Specifications

Chemical Name or Material Phosphorous-32
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.