Learn More
Strem, An Ascensus Company CAS . 100mg. (3aS8aS)-()-4488-Tetrakis(35-diethylphenyl)tetrahydro-22-dimethyl-6-phenyl-13-dioxolo[45-e]dioxaphosphepin. MFCD23705049
SDP
Supplier:  Strem, An Ascensus Company 151518100MG
Encompass_Preferred
100mg. (3aS8aS)-()-4488-Tetrakis(35-diethylphenyl)tetrahydro-22-dimethyl-6-phenyl-13-dioxolo[45-e]dioxaphosphepin. MFCD23705049. Molecular Weight 797.06. Molecular Formula C53H65O4P. Color/form off-white pwdr. Strem 15-1518. http//www.strem.com/catalog/v/15-1518/
Catalog No. 50-211-0753
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.