Learn More
Strem, An Ascensus Company CAS 149968-36-5. 250mg. 13-Bis(di-t-butylphosphinomethyl)benzene 99%. MFCD01630850
SDP
Supplier:  Strem, An Ascensus Company 150065250MG
Encompass_Preferred
CAS 149968-36-5. 250mg. 13-Bis(di-t-butylphosphinomethyl)benzene 99%. MFCD01630850. Molecular Weight 394.56. Molecular Formula C6H4[CH2P(C4H9)2]2. Color/form white pwdr. Strem 15-0065. http//www.strem.com/catalog/v/15-0065/
Catalog No. 50-211-0725
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.