Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phosphorus Pentabromide

Catalog No. 226734100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-673-4100G 100 g
22-673-425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 22-673-4100G Supplier Sigma Aldrich Supplier No. 226734100G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phosphorus pentabromide (PBr5) is a bromination agent generally used to convert alcohols to bromides and in the dibromination of ketones. PBr5 can also be used as the phosphorus source in the preparation of indium phosphide (InP) nanowires.

Specifications

CAS 7789-69-7
Quantity 100 g
Molecular Formula Br5P
Linear Formula PBr5
MDL Number MFCD00011437
Synonym Pentabromophosphorane
Molecular Weight (g/mol) 430.49 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.