Learn More
Sigma Organic Chemistry Phosphorus tribromide | 10G | 7789-60-8 | MFCD00011436 | >=99.99% trace metals basis
Supplier: Sigma Organic Chemistry 28846210G

Phosphorus tribromide, 10G
About This Item:
Synonym(s): Phosphorus(III) bromide
Linear Formula: PBr3
Molecular Weight: 270.69
CAS Number: 7789-60-8
MDL Number: MFCD00011436
UNSPSC Code: 12352300
Purity: >=99.99% trace metals basis
Boiling Point: 175 C (lit.)
Melting Point: -41.5 C (lit.)
Density: 2.88 g/mL at 20 C (lit.)
Refractive Index: n20/D 1.697 (lit.)
Storage: room temp
EINECS Number: 232-178-2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.