Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorylcholine Mouse anti-All, Clone: BH8, MilliporeSigma™

Catalog No. MABF208450 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2084-50 50 μL
MABF2084-200 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2084-50 Supplier MilliporeSigma Supplier No. MABF208450UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Phosphorylcholine, clone BH8 is a mouse monoclonal antibody that detects Phosphorylcholine and has been tested for use in ELISA, Flow Cytometry, Immunocytochemistry, Inhibition/Function studies, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphorylcholine
Applications ELISA, Flow Cytometry, Immunocytochemistry, Inhibition Assays, Western Blot
Classification Monoclonal
Clone BH8
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Host Species Mouse
Immunogen Bacteria (Streptococcus pneumoniae, non-encapsulated strain R36A).
Purification Method Purified by ion-exchange chromatography
Quantity 50 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone BH8 speifically detects phosphocholine conjugated to BSA.
Target Species All species
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.