Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phosphoserine Monoclonal Antibody (3C171)
GREENER_CHOICE

Catalog No. MA191608 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA191608 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA191608 Supplier Invitrogen™ Supplier No. MA191608
  • $669.00
    $550.65 / Each of 1

    Save $118.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA1-91608 detects Phosphoserine from rat samples.

Phosphorylated serine (phosphoserine) is an alpha amino acid that together with phosphothreonine, is one of the most ubiquitously modified amino acids in proteins. Many hormones, such as adrenaline (epinephrine), alter the activities of enzymes by stimulating the phosphorylation of serine and threonine, and these can have profound effects on cellular signal transduction, apoptosis and carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphoserine
Applications Dot Blot, ELISA, Western Blot
Classification Monoclonal
Clone 3C171
Concentration 4.4 mg/mL
Conjugate Unconjugated
Formulation Ascites with 15 mM sodium azide
Gene Alias C3H8NO6P; Dexfosfoserine; Fosforina; L-O-Phosphoserine; O-phospho-L-serine; O-Phosphoserine; Phospho-Serine; pSer
Host Species Mouse
Immunogen Phosphoserine conjugated to KLH.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.