Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phosphotyrosine Monoclonal Antibody (M375)
GREENER_CHOICE

Catalog No. PIMA547634
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547634 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547634 Supplier Invitrogen™ Supplier No. MA547634
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody detects phosphotyrosine-containing proteins on SDS-PAGE immunoblots of EGF treated A431 cells, as well as pervanadate treated A431, HeLa, Jurkat, and endothelial cells.

The role of tyrosine phosphorylation in transduction of the mitogenic signal from transmembrane receptors and in transformation by oncogene tyrosine kinases has been the subject of intense investigation for several years. While the phosphorylation of specific tyrosine residues has been shown to be a primary mechanism of signal transduction during normal mitogenesis, cell cycle progression and oncogenic transformation, its role in other areas such as differentiation and gap junction communication, is a matter of active and ongoing research. Antibodies that specifically recognize phosphorylated tyrosine residues have proved to be invaluable to the study of tyrosine -phosphorylated proteins and the biochemical pathways in which they function. The fluorescein (FITC) conjugate of clone PY20 anti-phosphotyrosine is especially useful for the detection of these P-Tyr proteins in immunohistochemical and immunocytochemical protocols in situations wherein the use of a secondary antibody would complicate detection of the protein(s) of interest.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphotyrosine
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone M375
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene NOS3
Gene Accession No. P29474, P70313, Q62600
Gene Alias Phosphotyrosine; pTyr; pY
Host Species Mouse
Immunogen Clone M375 was generated from tyrosine phosphorylated proteins purified from pervanadate treated A431 cells.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18127, 24600, 4846
Target Species Chemical, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.