Learn More
Invitrogen™ Phosphotyrosine Monoclonal Antibody (P-Tyr-01), Alexa Fluor™ 647
Shop All Thermo Scientific Products

Description
This antibody detects tyrosine phosphorylation in activated cells.
Specifications
Specifications
| Antigen | Phosphotyrosine |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | P-Tyr-01 |
| Concentration | 1 mg/mL |
| Conjugate | Alexa Fluor 647 |
| Formulation | PBS with 15mM sodium azide |
| Gene Alias | Phosphotyrosine; pTyr; pY |
| Host Species | Mouse |
| Immunogen | Phosphotyrosine conjugated to bovine serum albumin. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.