Learn More
BioVendor, LLC Phosphotyrosine Tie-1 Elisa BioVendor LLC
SDP
Supplier:  BioVendor, LLC REH010R
Encompass
The BioVendor Phosphotyrosine TIE-1 ELISA kit is a very rapid, convenient and sensitive assay kit that can monitor the activation or function of important biological pathways in cell lysates. This sandwich-based ELISA kit is an in vitro enzyme-linked immunosorbent assay for the measurement of Human phospho-TIE-1. An anti-TIE-1 antibody has been coated onto a 96-well plate. Samples are pipetted into the wells and phosphorylated and unphosphorylated TIE-1 present in a sample is bound to the wells by the immobilized antibody. The wells are washed and biotinylated antiphosphotyrosine antibody is used to detect only tyrosine-phosphorylated protein. After washing away unbound antibody, HRP-conjugated streptavidin is pipetted to the wells. The wells are again washed, a TMB substrate solution is added to the wells and color develops in proportion to the amount of phospho-TIE-1 bound. The Stop Solution changes the color from blue to yellow, and the intensity of the color is measured at 450 nm.
Catalog No. 50-240-6387
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.