Learn More
Advanced Biomatrix PhotoHA-Soft Methacrylated Hyaluronic Acid (GM-HA) with Ruthenium Kit
Supplier:  Advanced Biomatrix 54051KIT
Encompass_Preferred
The PhotoHA Soft kit consists of purified methacrylated hyaluronic acid and a Ruthenium photoinitiator. PhotoHA is 100 mg of lyophilized methacrylated hyaluronic acid (GM-HA). The photoinitiator consists of Ruthenium and Sodium Persulfate which need to be formulated in 1X PBS or cell culture media, allowing for visible light photocrosslinking of the HA at 400-450 nm. PhotoHA provides 3D hyaluronic acid hydrogels with the unique attributes to be prepared at various concentrations and photocrosslinked to provide various gel stiffness. GM-HA forms softer hydrogels than HAMA. These hydrogels provide native-like 3D HA gels with the unique attributes to be prepared at various concentrations and photocrosslinked to provide various gel stiffness. Hyaluronic acid is the most abundant glycosaminoglycan in the body being an important component of several tissues throughout the body.
Catalog No. NC2368625
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.