Learn More
POZZETTA PRODUCTS PHOTOMASK,ANTI-STATIC,0.12,5/P
Supplier:  POZZETTA PRODUCTS PM600NP12
Encompass
Anti-Static Polypropylene. This 6-inch translucentmask package offers the durability and impact strength of polypropylene. Cleanroom manufactured and packaged in cleanroom bags.Clear anti-static polypropylene. Standard stock for 0.120″ thick.
Catalog No. NC1860817
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.