Learn More
Advanced Biomatrix PhotoSericin, Methacrylated Sericine, 1 gram with LAP kit
Supplier:  Advanced Biomatrix 54311KIT
Encompass_Preferred
PhotoSericin with LAP kit is 1 gram of lyophilized, sterile, methacrylated sericin with 100 mg LAP. PhotoSericin provides 3D hydrogels with the unique attributes to be prepared at various concentrations and photocrosslinked (requires a photoinitiator) to provide various gel stiffness. PhotoSericin is a highly versatile solution designed for researchers and scientists seeking advanced materials for tissue engineering, drug delivery, and regenerative medicine applications. PhotoSericin provides 3D tunable gels with the unique attributes to be prepared at various concentrations and crosslinked to provide various gel stiffnesses. The photoinitiator consists of LAP which allows visible light crosslinking of the sericin at 405 nm. LAP is provided separately as a powder to provide customization and precise control over the crosslinking process.
Catalog No. NC2410849
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.