Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHOX2A Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735076 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5076 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5076 Supplier Proteintech Group Inc Supplier No. 258041AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PHOX2A and PHOX2B (Paired mesoderm homeobox protein) are closely related, paired-homeodomain transcription factors that function as determits of the noradrenergic phenotype during embryogenesis. PHOX2 proteins are crucial for the regulation of endogenous hydroxylases in neural crest cells and promote sympathetic neuron generation. Human PHOX2B contains one DNA binding homeobox domain and is required for the differentiation of all central and nonperipheral noradrenergic centers in the brain. In contrast, PHOX2A controls only the differentiation of the main noradrenergic center of the brain. Regulation of PHOX2 may have therapeutic utility in aging or disorders involving degeneration of noradrenergic neurons.

Specifications

Antigen PHOX2A
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PHOX2A
Gene Accession No. O14813
Gene Alias ARIX, ARIX1 homeodomain protein, CFEOM2, FEOM2, paired like homeobox 2a, PHOX2A, PMX2A
Gene Symbols PHOX2A
Host Species Rabbit
Immunogen PHOX2A Fusion Protein Ag22873
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.