Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHRF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554103
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554103 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554103 Supplier Invitrogen™ Supplier No. PA554103
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SCIPSVLKPV EPSLGLLRAD IGAASLSLFG DPYELDPFDS SEELSANPLS PLSAKRRALS RSALQSHQPV ARPVSVGLSR RRLPAAVPEP DLEEEPVPDL LGSILSGQSL LMLGSSDVII HRDGSLSAKR AAPVSFQR Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 80%.

Plays important roles in controlling the metabolism of fatty acids at differents levels. Acts as a magnesium-dependent phosphatidate phosphatase enzyme which catalyzes the conversion of phosphatidic acid to diacylglycerol during triglyceride, phosphatidylcholine and phosphatidylethanolamine biosynthesis in the reticulum endoplasmic membrane. Acts also as a nuclear transcriptional coactivator for PPARGC1A to modulate lipid metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHRF1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHRF1
Gene Accession No. Q9P1Y6
Gene Alias AA673488; CTD-binding SR-like protein rA9; Kiaa1542; PHD and RING finger domain-containing protein 1; PHD and ring finger domains 1; phrf1; PPP1R125; protein phosphatase 1, regulatory subunit 125; RNF221
Gene Symbols PHRF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHRF1. Recombinant protein control fragment (Product #RP-91599).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57661
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.