Learn More
Sartorius pHrodo Red Cell Labeling Kit for Phagocytosis Assays, Real-Time Analysis, pH-Sensitive Fluorophore, 25.125 units
Catalog No. NC2017730 Shop All Sartorius Products
Encompass
The Sartorius pHrodo Red Cell Labeling Kit for Incucyte Phagocytosis Assays provides a streamlined solution for quantitative analysis of phagocytic activity in live-cell imaging. Utilizing a unique pH-sensitive fluorophore, this kit enhances detection of internalized cells without the need for wash steps or quenching reagents, making it ideal for various applications in cell biology. Its compatibility with Incucyte systems and high labeling capacity make it a versatile tool for researchers in phagocytosis studies.
Catalog No. NC2017730 Supplier Sartorius Supplier No. 4649
The Sartorius pHrodo Red Cell Labeling Kit for Incucyte Phagocytosis Assays provides a streamlined solution for quantitative analysis of phagocytic activity in live-cell imaging. Utilizing a unique pH-sensitive fluorophore, this kit enhances detection of internalized cells without the need for wash steps or quenching reagents, making it ideal for various applications in cell biology. Its compatibility with Incucyte systems and high labeling capacity make it a versatile tool for researchers in phagocytosis studies.