Learn More
Sartorius IncuCyte pHrodo Red Zymosan Bioparticles for Phagocytosis
Catalog No. NC1665761 Shop All Sartorius Products
Encompass
One vial of 1 mg sterile lyophilized solid sufficient for 100-200 live-cell imaging phagocytosis tests in 96-well format
Catalog No. NC1665761 Supplier Sartorius Supplier No. 4617
One vial of 1 mg sterile lyophilized solid sufficient for 100-200 live-cell imaging phagocytosis tests in 96-well format