Learn More
Clontech Labs 3P PHSG 298 DNA 25 UG
SDP
Supplier:  Clontech Labs 3P 3298
Encompass
Vector; DNA; Takara; pHSG 298; karl pUC vector; 25ug; 2,686bp; 250-1000ug/mL; pUC 18 MCS
Catalog No. 50-443-746
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.