Learn More
Clontech Labs 3P PHSG 299 DNA 25 UG
Supplier: Clontech Labs 3P 3299
Vector, DNA; Takara; pHSG 299; karl pUC vector; 25ug; 2,676bp; 250-1000ug/mL; pUC19-like MCS
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.