Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PHT 427

Catalog No. 459850
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-985-0 50 mg
45-981-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-985-0 Supplier Tocris Bioscience™ Supplier No. 4598/50
This item is not returnable. View return policy

Dual Akt and PDK1 inhibitor; antitumor

PHT 427 is a dual Akt and PDK1 inhibitor; binds the pleckstrin homology (PH) domain of Akt and PDK1 (Ki values are 2.7 and 5.2 μM, respectively). Inhibits Akt phosphorylation and Akt downstream targets; also induces apoptosis in vitro. Displays synergistic antitumor effect when combined with chemotherapeutics in breast cancer mouse models.

Specifications

Purity >98%
Quantity 50 mg
Product Type PHT 427
Molecular Weight (g/mol) 409.61
Inhibitors PHT 427

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.