Learn More
Aqua Solutions Phthalate-Phosphate Reagent Powder Pillows, 25 mL, pk/50
SDP
Supplier:  Aqua Solutions HA2150166
Encompass
For low-level determination of nickel and cobalt by the PAN method. Hach method 8078. Sample Size: 25 mL. Pack of 50 Powder Pillows.
Catalog No. NC2122118
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.