Learn More
Medchemexpress LLC Phthalic acid anhydride-d4 | 75935-32-9 | MFCD00145119 | 98.0% | 152.14 | C8D4O3 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW019814500MG
Encompass_Preferred
Phthalic acid anhydride-d4 is the deuterium-labeled form of phthalic anhydride, supplied as an off-white to gray solid for use as an isotopic standard and synthetic reagent in analytical and preparative chemistry. It is commonly used in mass spectrometry, NMR, tracer studies, and as a labeled intermediate for synthesis.
  • Deuterium-labeled reagent for isotopic tracing and mass spectrometry.
  • High isotopic enrichment (98 atom % D) for distinct mass shifts.
  • Off-white to gray solid with recommended low-temperature storage for stability.
  • Suitable as an internal standard, tracer, or synthetic intermediate.
  • Molecular formula C8D4O3 and molecular weight 152.14 aid identification and calculations.
Catalog No. 50-002-99584