Learn More
Sigma Organic Chemistry Phthalic acid dipotassi | 25G | 4409-98-7 | MFCD00040595
Supplier: Sigma Organic Chemistry 37648525G

Phthalic acid dipotassi , 25G
About this item:
CAS #: 4409-98-7
MDL #: MFCD00040595
Purity: ≥98
Chemical Formula: C8H4K2O4
Molecular Weight: 242.31
UNSPSC Code: 12352100
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.