Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalic Anhydride, ACS, 99.0-100.2%, Spectrum™ Chemical
SDP

Catalog No. 18612364 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-364 25 g Poly Bottle
18-612-365 500 g Poly Bottle
18-612-367 2.5 kg Poly Bottle
18-612-366 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-612-364 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P114025GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85-44-9
Molecular Formula C8H4O3
Molecular Weight (g/mol) 148.12
InChI Key LGRFSURHDFAFJT-UHFFFAOYSA-N
IUPAC Name 1,3-dihydro-2-benzofuran-1,3-dione
SMILES O=C1OC(=O)C2=CC=CC=C12

Specifications

CAS Max % 1
Melting Point Within 3°C
Ignition Residue 0.0001
Packaging Poly Bottle
Quantity 25 g
Percent Purity 99 to 100.2%
Grade ACS

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.