Learn More
Sigma Aldrich Fine Chemicals Biosciences fmoc Lysbiotin OH Novabioc
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 8520978500
Encompass
A modified lysine derivative for the preparation of biotin-labeled peptides by fmoc SPPS.The product number for this product was previously 04-12-1237.To obtain a certificate of analysis (CoA) of a lot that begins with the letter A please select the option in the right hand menu Request a COA for Lots starting with A.
Catalog No. 50-289-7425
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.