Learn More
Medchemexpress LLC HY-W015820 500mg Medchemexpress, Phthalide CAS:87-41-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW015820500MG
Encompass_Preferred
Medchemexpress, HY-W015820 500mg Phthalide CAS:87-41-2 Phthalide is a promising chemical scaffold with a potent anti-inflammatory efficacy. Phthalide can be used to synthesize a variety of phthalide derivatives including anti-inflammatory agent, antimicrobial, antioxidant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3385
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.