Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalylsulfacetamide, MP Biomedicals™

Catalog No. ICN10489380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10489380 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10489380 Supplier MP Biomedicals, Inc Supplier No. 0210489380

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Slightly soluble in water

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-69 , 7681-11-0
Molecular Formula C16H14N2O6S
Synonym 2-([(4-([Acetylamino]sulfonyl)phenyl)amino]carbonyl)benzoic acid

Specifications

Color White
Quantity 100 g
Formula Weight 362.4
Physical Form Powder
Chemical Name or Material Phthalylsulfacetamide
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.