Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phusion™ Hot Start II DNA Polymerases (2 U/μL)

Catalog No. f549s
Change view
Click to view available options
Color:
Colorless
Green
No. of Reactions:
100 Reactions
2000 Reactions
500 Reactions
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. of Reactions
F549S Colorless 100 Reactions
F537S Green 100 Reactions
F537L Green 500 Reactions
F549L Colorless 500 Reactions
F549XL Colorless 2000 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. F549S Supplier Thermo Scientific™ Supplier No. F549S
Only null left
Add to Cart
Add to Cart

Includes

Phusion Hot Start II High–Fidelity DNA Polymerase

  • Phusion Hot Start II High–Fidelity DNA Polymerase (2U/μL)
  • 5x Phusion HF Buffer
  • 5x Phusion GC Buffer
  • DMSO
  • 50mM MgCl2 solution
Phusion Green Hot Start II High–Fidelity DNA Polymerase
  • Phusion Hot Start II High–Fidelity DNA Polymerase (2U/Glc muL)
  • 5x Phusion Green HF Buffer
  • 5x Phusion Green GC Buffer
  • DMSO
  • 50mM MgCl2 solution
Both Phusion and Phusion Green Buffer provide 1.5 mM MgCl2 in the final 1x concentration

Thermo Scientific Phusion High-Fidelity DNA polymerases set a gold standard for high performance PCR.

Thermo Scientific Phusion High-Fidelity DNA polymerases set a gold standard for high performance PCR. Featuring an error rate 50-fold lower than that of Taq and 6-fold lower than that of Pfu, Phusion High-Fidelity DNA Polymerase is excellent choice for cloning and other applications requiring high fidelity. Phusion DNA polymerases offer robust performance with short protocol times, even in the presence of PCR inhibitors, and generate higher yields with lower enzyme amounts than other DNA polymerase.

With Phusion Hot Start II High-Fidelity DNA Polymerase amplification proceeds without the production of nonspecific products due to the combination of Phusion DNA Polymerase and a reversibly bound, specific Affibody ligand that inhibits DNA polymerase activity at room temperature. The Affibody ligand also inhibits the 3' to 5' exonuclease activity of the polymerase, thus preventing degradation of primers and template DNA during reaction set up. At temperatures that promote polymerase activity, the ligand is released, rendering the polymerase fully active. Phusion Hot Start II DNA Polymerase is immediately reactivated at high temperatures, so it does not require a separate activation step in PCR protocols.

Phusion Hot Start II High-Fidelity DNA Polymerase is a combination of Phusion Hot Start II DNA Polymerase and the 5X Phusion buffers. The buffers include a density reagent and two tracking dyes which do not interfere with the robust performance of Phusion Hot Start II DNA Polymerase. Researchers may directly load PCR products on a gel making this a versatile option for use in downstream applications such as DNA sequencing, ligation and restriction digestion.

Highlights

  • Reaction set up at room temperature
  • No non-specific amplification and primer degradation during reaction set up
  • Zero-time reactivation due to unique hot start technology
  • High fidelity (52X Taq)
  • Fast PCR due to short extension times (15–30 s/kb)
  • Robust reactions, minimal optimization needed
  • Increased product yields with minimal enzyme amounts
  • Direct loading on gels

Applications

  • High-fidelity PCR
  • High throughput
  • Amplification of difficult (GC-rich) templates
  • Template generation for sequencing
  • Multiplex PCR
  • Long-range PCR
  • Cloning
  • Mutagenesis
  • Microarray

Using Phusion DNA polymerases

Annealing rules for Phusion DNA polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 U/μL
Content And Storage • Phusion Hot Start II High-Fidelity DNA Polymerase (2 U/μL)
• 5X Phusion HF & GC Buffers
• DMSO
• 50 mM MgCl2 solution

Phusion HF Buffer and Phusion GC Buffers each provide 1.5 mM MgCl2 in the final 1X concentration.
Store at -20°C.
GC-Rich PCR Performance High
Polymerase Phusion Hot Start II DNA Polymerase
Reaction Speed Fast
Product Type Hot Start DNA Polymerase
Quantity 100 units
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq) 52X
Hot Start Built-In Hot Start
No. of Reactions 100 Reactions
Overhang Blunt
Reaction Format Standalone
Size (Final Product) 20 kb or less
Color Colorless
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.