Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phusion™ Plus PCR Master Mixes

Catalog No. F631L
Change view
Click to view available options
Color:
Colorless
Green
No. of Reactions:
100 Reactions
2000 Reactions
500 Reactions
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. of Reactions
F631L Colorless 500 Reactions
F632L Green 500 Reactions
F632S Green 100 Reactions
F632XL Green 2000 Reactions
F631S Colorless 100 Reactions
F631XL Colorless 2000 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. F631L Supplier Thermo Scientific™ Supplier No. F631L
Only null left
Add to Cart
Edge
Add to Cart

Thermo Scientific Phusion Plus PCR Master Mix is a convenient, ready-to-use 2X mixture of Phusion Plus DNA Polymerase, buffer, magnesium chloride, and dNTPs.

Thermo Scientific Phusion Plus Green PCR Master Mix is a convenient, ready-to-use 2X mixture of Phusion Plus DNA Polymerase, buffer, magnesium chloride, and dNTPs. The master mix format simplifies reaction setup with minimal pipetting steps and retains all features of Phusion Plus DNA Polymerase. Additionally, two tracking dyes and a density reagent are pre-added to the master mix for direct loading of PCR products on gels.

Features of Phusion™ Plus PCR Master Mix include:

  • High fidelity—provides >100X higher sequence accuracy than Taq polymerase
  • Convenient setup—enables a universal annealing temperature of 60°C in PCR
  • Enhanced specificity—hot-start mechanism reduces non-specific amplification and primer degradation
  • Benchtop stability—automation enabled due to 24-hour stability of assembled reactions
  • High inhibitor tolerance—works with DNA of suboptimal purity

Phusion Plus DNA Polymerase is a hot-start, high-fidelity DNA polymerase that brings together protein fusion technology and universal primer annealing. It is a Pyrococcus-like proofreading DNA polymerase fused to a processivity-enhancing domain, enabling Phusion Plus PCR Master Mix to generate PCR sequences with high accuracy, sensitivity, and inhibitor tolerance. Its novel reaction buffer includes proprietary additives that stabilize primer/template duplexes during the annealing step at 60°C. This unique buffer formulation increases PCR specificity without the need to optimize annealing temperature for each primer pair, helping save time and avoid mistakes in PCR runs. Furthermore, with Phusion Plus PCR Master Mix and its universal annealing feature, different PCR reactions can be cycled together using the same protocol.

Phusion Plus PCR Master Mix is inactive at ambient temperatures due to its hot-start mechanism mediated by Affibody molecules, allowing setup and storage of assembled PCR reactions at room temperature. Enzyme activity is fully restored after the initial denaturation step.

Applications

  • High-fidelity PCR
  • Cloning
  • Template generation for sequencing
  • Amplification of difficult (GC-rich) templates
  • Long-range PCR (up to 20 kb)
  • Mutagenesis
  • High-throughput PCR
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • 2X Phusion Plus PCR Master Mix (10 x 1.25 mL)
• 5X Phusion GC Enhancer (4 x 1.25 mL)
• Nuclease-free water (10 x 1.25 mL)

Store at –15°C to –25°C
GC-Rich PCR Performance High
Polymerase Phusion Plus DNA Polymerase
Reaction Speed 15 to 30 sec./kb
Quantity 500 x 50 μL reactions
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq) >100X
Hot Start Built-In Hot Start
No. of Reactions 500 Reactions
Overhang Blunt
Reaction Format Ready to use master mix
Size (Final Product) 20 kb or less
Color Colorless
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.