Learn More
Sigma Aldrich Fine Chemicals Biosciences PhytagelTM BioReagent suit5KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P81695KG
Encompass
Typical working concentration 1.5-2.5 g/L in plant tissue culture media up to 10 g/L in microbiological media. Phytagel(TM) requires the presence of cations (especially divalent) for gelling to occur. Concentrations of calcium and magnesium contained in most plant tissue culture media are typically sufficient for gelation. Low-salt media formulations especially those used in microbiological applications may require supplementation with additional calcium or magnesium salts (e.g. CaCl2 or MgSO4) or higher concentrations of Phytagel.
Catalog No. 50-290-7542
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.