Learn More
Apexbio Technology LLC Nafarelin Acetate 76932-56-4 10mg
Supplier: Apexbio Technology LLC C644410
Nafarelin Acetate is a synthetic agonist targeting gonadotropin-releasing hormone (GnRH) receptors It is designed to modulate the hypothalamic-pituitary-gonadal axis by initially stimulating and subsequently downregulating GnRH receptors in the pituitary gland thereby suppressing secretion of luteinizing hormone (LH) and follicle-stimulating hormone (FSH) Nafarelin Acetate exerts its biological activity primarily through receptor downregulation and suppression of pituitary hormone release Based on these pharmacological properties Nafarelin Acetate holds research potential in the investigation of endocrine signaling pathways associated with reproductive development disorders including central precocious puberty and endometriosis
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.