Learn More
Sigma Aldrich Fine Chemicals Biosciences Phytic acid solution 50 w1L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 5936481L
Encompass
Phytic acid is a mineral chelator that can bind to minerals to form mineral-phytate complex.It can also complex with metal ions bound to the surface of magnesium alloy to form a conversion coating which can improve the resistance of magnesium alloy towards corrosion.
Catalog No. 50-289-7492
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.