Learn More
Biotage PHYTIP COLUMNS W80UL IMAC
SDP
Supplier:  Biotage PTR918003
Encompass
Box of 96 PhyTip columns (1 ml volume) containing 80 ul of IMAC affinity resin per column
Catalog No. NC1798100
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.