Learn More
CPC Scientific H-gamma-Glu-Cys-gamma-Glu-Cys-Gly-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PHYT003A
Encompass
A glutathione-derived heavy metal-detoxifying peptide of higher plants consisting of 2 units of gammaGlu-Cys.SEQUENCE: H-gamma-Glu-Cys-gamma-Glu-Cys-Gly-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: (gammaE-C)2-GMOLECULAR FORMULA: C18H29N5O10S2MOLECULAR WEIGHT: 539.58STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: H--Glu-Cys--Glu-Cys-Gly-OHRESEARCH AREA: MiscellaneousREFERENCES:Grill, E. et al. Science 230, 674 (1985)Rauser, WE. Plant Physiol. 109, 1141 (1995)SKU(s): PHYT-003A
Catalog No. 50-211-0151
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.