Learn More
Medchemexpress LLC HY-N7038 10mg Medchemexpress, Phytohemagglutinin CAS:9008-97-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN703810MG
Encompass_Preferred
Medchemexpress, HY-N7038 10mg Phytohemagglutinin CAS:9008-97-3 Phytohemagglutinin (PHA-M) is a lectin, obtained from the red kidney bean that binds to the membranes of T-cells, stimulates metabolic activity, cell division, and involves inflammatory pathways. Phytohemagglutinin (PHA-M) induces apoptosis via increasing proapoptotic protein Bax and activating caspases-3[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0491
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.