Learn More
Medchemexpress LLC Phytosterols | 949109-75-5 | 98.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12232010MG
Encompass_Preferred
Phytosterols are plant-derived sterols supplied as a powder for research use in studies of cardiovascular disease and cancer. The material is identified by CAS 949109-75-5 and has an HPLC assay of 98.0%. Recommended storage: powder at -20°C for long-term storage or 4°C for shorter-term; in solvent store at -80°C or -20°C depending on duration.
  • 98.0% purity by HPLC.
  • Powder form suitable for biochemical and pharmacological assays.
  • Used in cardiovascular disease and cancer research.
  • Stable when stored at -20°C for long-term preservation.
  • Provided in small research quantities and packaged for laboratory use.
Catalog No. 50-002-31281