Learn More
Echelon Biosciences Research Labs PI(3 5)P2 DiC4 500 ug
SDP
Supplier:  Echelon Biosciences Research Labs P3504500UG
Encompass
PI(3,5)P2 diC4 (Phosphatidylinositol 3,5-bisphosphate diC4) is a synthetic, purified dibutanoyl PI(3,5)P2.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3,5-bisphosphate is in very low abundance and is required for retrograde membrane trafficking from lysosomal and late endosomal compartments to the Golgi and is involved in autophagy. PI(3,5)P2 levels are controlled by the 5-kinase PIKfyve and the 5-phosphatase Fig4/Sac3 and disregulation is linked to several human neuropathies (eg Chacot-Marie-Tooth disease). This item is non returnable
Catalog No. 50-720-583
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.