Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Echelon Biosciences Research Labs AS605240 (PI3-K gamma Inhibitor 1) 1 mg
SDP

Supplier:  Echelon Biosciences Research Labs B03015MG

Encompass

5-quinoxilin-6-methylene-1,3-thiazolidine-2,4-dione (AS605240) is an ATP-competitive inhibitor of PI3-K (IC50 = 8 nM). It is selective for the -isoform (IC50: PI 3-K = 60 nM, PI 3-K = 270 nM, PI 3-K = 300 nM) and shows no notable activity against a wide array of protein kinases at 1 M. In addition, AS605240 suppresses the progression of joint inflammation in a mouse model of rheumatoid arthritis and suppresses inflammation in a mouse model of lupus.nn


nn

Solubility

nwater: insoluble, DMSO: 2.5 mg/mLn

Storage

nstore dry between 2-8oC. Stock solutions should be stored frozen (-20oC).n

References

n1) M. Camps et al. "Blockade of PI3Kg suppresses joint inflammation and damage in mouse models of rheumatoid arthritis" This item is non returnable

Catalog No. 50-720-013


May include imposed supplier surcharges.
Only null left
Add to Cart
This item is not returnable. View return policy

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.