Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI 3065, >98%, Tocris Bioscience™

Catalog No. 591910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-191-0 10 mg
59-195-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 59-191-0 Supplier R&D Systems Supplier No. 5919/10
This item is not returnable. View return policy

Potent and selective PI 3-kinase p110δ inhibitor

Potent and selective PI 3-kinase p110δ inhibitor (IC50 = 5 nM; Ki = 1.5 nM). Exhibits >100-fold selectivity over p100α, p100γ and p100β isoforms (IC50 values are 600, 910, >10,000 nM, respectively). Attenuates 4TI tumor growth and metastasis in vivo.

Chemical Identifiers

CAS 955977-50-1
Molecular Formula C27H31FN6OS
Molecular Weight (g/mol) 506.64
MDL Number MFCD28167771
InChI Key YDNOHCOYQVZOMC-UHFFFAOYSA-N
Synonym 4-6-4-cyclopropylmethyl piperazin-1-yl methyl-2-5-fluoro-1h-indol-4-yl thieno 3,2-d pyrimidin-4-yl morpholine, d01ssi, 4-6-4-cyclopropylmethyl piperazin-1-yl methyl-2-5-fluoro-1h-indol-4-yl thieno 3,2, 6-4-cyclopropylmethyl-piperazin-1-ylmethyl-2-5-fluoro-1h-indol-4-yl-4-morpholin-4-yl-thieno 3,2-d pyrimidine, 6-4-cyclopropylmethyl-1-piperazinyl methyl-2-5-fluoro-1h-indol-4-yl-4-4-morpholinyl-thieno 3,2-d pyrimidine
PubChem CID 24937012
IUPAC Name 4-(6-{[4-(cyclopropylmethyl)piperazin-1-yl]methyl}-4-(morpholin-4-yl)thieno[3,2-d]pyrimidin-2-yl)-5-fluoro-1H-indole
SMILES FC1=CC=C2NC=CC2=C1C1=NC(N2CCOCC2)=C2SC(CN3CCN(CC4CC4)CC3)=CC2=N1

Specifications

Linear Formula C27H31FN6OS
Quantity 10 mg
Solubility Information Soluble to 20mM in DMSO
Formula Weight 506.64
Chemical Name or Material 6-[[4-(Cyclopropylmethyl)-1-piperazinyl]methyl]-2-(5-fluoro-1H-indol-4-yl)-4-(4-morpholinyl)-thieno[
Grade >98%
Recommended Storage Store at −20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.