Learn More
Echelon Biosciences Research Labs PI(4)P DiC4 1 mg
SDP
Supplier:  Echelon Biosciences Research Labs P40041MG
Encompass
Phosphatidylinositol 4-phosphate diC4 (PI(4)P diC4) is a synthetic, purified dibutanoyl PI(4)P.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 4-phosphate (PI(4)P) is the biosynthetic precursor to PI(4,5)P2 and has an important roles in regulating sphingomyelin and glycosphingolipid metabolism and membrane trafficking at the exit of the Golgi complex. This item is non returnable
Catalog No. 50-720-616
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.