Learn More
ProteinSimple iCE280/iCE3 pI Marker 9.22 200ul
Supplier:  ProteinSimple 102231
Encompass
9.2 pi Value Marker for pI determination that will give you both consistent and accurate measurement of isoelectric point on your iCE system. It can be used for whole-column and single-point capillary isoelectric focusing (cIEF) detection to measure your sample protein's isoelectric point, and for quality control of sample analysis. You'll get 200 L of pI marker stock solution in every vial.
Catalog No. NC9662400
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.