Learn More
Sigma Aldrich Fine Chemicals Biosciences PI3 Kinase (p110α/p85α) Protein
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 14602
Encompass
PI3 Kinase (p110α/p85α) Protein | Active complex of N-terminal His6-tagged recombinant full-length human p110α & untagged, recombinant, full length, human p85α. For use in Kinase Assays, 20ug
Catalog No. NC2740067