Learn More
MilliporeSigma™ PI3 Kinase Activity/Inhibitor ELISA
Catalog No. 17493 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
96-well Plate
Catalog No. Quantity
17-493 96-well Plate
1 options
Catalog No. 17-493 Supplier MilliporeSigma™ Supplier No. 17493
Add to Cart
Add to Cart
Includes
  • Glutathione-Coated Plate, 96-well strip format
  • 20% Tween™ 20
  • 20X TBS Wash
  • Substrate TMB
  • Stop Buffer
  • SA-HRP Detection Reagent
  • GRP1-GST
  • Kinase Reaction Buffer, 5X
  • Wortmannin
  • PIP2
  • Biotinylated-PIP3
  • PI3K, p110α
  • PI3K, p110β
  • PI3K, p110δ
  • PI3K, p120γ
Used to measure & quantify PI 3-Kinase levels
  • Used to measure and quantify PI 3-Kinase levels
  • Applications: Kinase Assay
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Kinase Assay
Quantity 96-well Plate