Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI3K p85 alpha Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4G3C11, Proteintech
SDP

Catalog No. 501736326 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6326 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6326 Supplier Proteintech Group Inc Supplier No. 602251IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Phosphatidylinositol 3-kinase phosphorylates the inositol ring of phosphatidylinositol at the 3-prime position. The enzyme comprises a 110 kDa catalytic subunit and a regulatory subunit of either 85, 55, or 50 kDa. It binds to activated (phosphorylated) protein-Tyr kinases, through its SH2 domain, and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane. Phosphatidylinositol 3-kinase plays an important role in the metabolic actions of insulin, and a mutation in this gene has been associated with insulin resistance. During insulin stimulation, it also binds to IRS-1.

Specifications

Antigen PI3K p85 alpha
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4G3C11
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PIK3R1
Gene Accession No. P26450, P27986, Q63787
Gene Alias GRB1, p85, p85 ALPHA, PI 3 Kinase p85 alpha, PI 3-kinase p85alpha, PI3 kinase subunit p85 alpha, PI3K p85(alpha), PI3K regulatory subunit alpha, PIK3R1
Gene Symbols PIK3R1
Host Species Mouse
Immunogen PI3K p85 alpha Fusion Protein Ag2344
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100623707, 18708, 25513, 5295
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.