Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PI4KA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528570 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528570 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528570 Supplier Invitrogen™ Supplier No. PA528570
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (86%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PI4KB and PI4KA are the two human type III PI4 kinases, which share sequence similarly with PI3 kinases and phosphorylate the 4 position of phosphotidyl inositol. The mammalian PI 4-kinases have been classified into two types, II and III, based on their molecular mass, and modulation by detergent and adenosine. The protein encoded by PI4KA is a type III enzyme that is not inhibited by adenosine. Two transcript variants encoding different isoforms have been described for PI4KA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI4KA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene PI4KA
Gene Accession No. P42356
Gene Alias leuserpin 2; phosphatidylinositol 4-kinase; phosphatidylinositol 4-kinase 230; phosphatidylinositol 4-kinase a; phosphatidylinositol 4-kinase alpha; phosphatidylinositol 4-kinase, catalytic, alpha; phosphatidylinositol 4-kinase, catalytic, alpha polypeptide; phosphatidylinositol 4-kinase, type III, alpha; pi4K230; Pi4ka; PI4K-ALPHA; PI4-kinase alpha; Pik4; PIK4CA; PMGYCHA; ptdIns-4-kinase alpha; testicular secretory protein Li 35
Gene Symbols PI4KA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PI4 kinase alpha. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5297
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.