Learn More
Description
These sequences are 95% and 100% homologous in mouse and rat. Suggested positive control: liver, Jurkat. Immunogen sequence: KKIKELYETR YAKKNC & YSDTSCPQED QY.
Specifications
Specifications
| Antigen | PIASy |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | PIAS4 |
| Gene Accession No. | Q8N2W9, Q9JM05 |
| Gene Alias | e3 sumo- ligase pias4; E3 SUMO-protein ligase PIAS4; FLJ12419; MGC35296; Pias4; Piasg; Pias-gamma; PIASy; protein inhibitor of activated STAT 4; protein inhibitor of activated STAT PIASy; protein inhibitor of activated STAT protein 4; Protein inhibitor of activated STAT protein gamma; protein inhibitor of activated STAT protein PIASy; protein inhibitor of activated STAT, 4; RING-type E3 ubiquitin transferase PIAS4; zinc finger, MIZ-type containing 6; ZMIZ6 |
| Gene Symbols | PIAS4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
