Learn More
Abcam Piceatannol, Syk Tyrosine Kinase Inhibitor, >99% Purity, 50 mg
Description
- Molecular Weight: 244.25 g/mol
- Purity: >99%
- Potency: Syk tyrosine kinase inhibitor (Ki = 15 μM at Syk)
- Biological Activity: Exhibits anti-inflammatory, antiproliferative, and immunomodulatory effects
- Form: Solid
- CAS Number: 10083-24-6
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Solid |
| Content And Storage | Store at 4°C. |
| Product Type | Antioxidant |
| Buffer | 100% Piceatannol |
| Shipping Condition | Ambient |
| Target Molecule | Syk Tyrosine Kinase |
| Formulation | 100% Piceatannol |
| Quantity | 50 mg |
| For Use With (Application) | Syk tyrosine kinase inhibition |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.