Learn More
Abcam Piceatannol, Syk Tyrosine Kinase Inhibitor, Solid, 10 mg
Description
- Activity: Syk tyrosine kinase inhibitor (Ki = 15 μM at Syk)
- Purity: >99%
- Molecular Weight: 244.25 g/mol
- Form: Solid
- Biological Activities: Exhibits anti-inflammatory, antiproliferative and immunomodulatory effects
- Storage: Store at +4°C
- Shipping: Ambient - Can Ship with Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | Syk Tyrosine Kinase |
| Form | Solid |
| Shipping Condition | Ambient |
| Buffer | 100% Piceatannol |
| Content And Storage | Store at 4°C. |
| Formulation | 100% Piceatannol |
| Quantity | 10 mg |
| For Use With (Application) | Syk tyrosine kinase inhibition |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.