Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Piceatannol
SDP

Catalog No. 155410 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-541-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-541-0 Supplier Tocris Bioscience™ Supplier No. 1554/10

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibits TNF-induced NF-κB activation

Anti-inflammatory, immunomodulatory and antiproliferative agent. Inhibits p56lck and syk protein tyrosine kinases and inhibits TNF-induced NF-κB activation and gene expression. Synthesis results from conversion of resveratrol (Cat. No. 1418) by cytochrome P450 1B1.

Chemical Identifiers

CAS 10083-24-6
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
InChI Key CDRPUGZCRXZLFL-OWOJBTEDSA-N
Synonym piceatannol, 3-hydroxyresveratol, astringinin, piceatanol, e-4-3,5-dihydroxystyryl benzene-1,2-diol, 3,3',4'5-tetrahydroxystilbene, 3,5,3',4'-tetrahydroxystilbene, 4-e-2-3,5-dihydroxyphenyl ethenyl benzene-1,2-diol, 3,3',4,5'-tetrahydroxystilbene, unii-6ks3ls0d4f
PubChem CID 667639
ChEBI CHEBI:28814
IUPAC Name 4-[(E)-2-(3,5-dihydroxyphenyl)ethenyl]benzene-1,2-diol
SMILES C1=CC(=C(C=C1C=CC2=CC(=CC(=C2)O)O)O)O

Specifications

Quantity 10 mg
Formula Weight 244.25
Percent Purity >98%
Chemical Name or Material Piceatannol
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.